Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human AKAP9 Partial ORF (NP_671700, 3812 a.a. - 3911 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00010142Q01S

 View more versions of this product

Catalog No. 89-932-997


Add to cart

Description

Description

The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. Alternate splicing of this gene results in at least two isoforms that localize to the centrosome and the Golgi apparatus, and interact with numerous signaling proteins from multiple signal transduction pathways. These signaling proteins include type II protein kinase A, serine/threonine kinase protein kinase N, protein phosphatase 1, protein phosphatase 2a, protein kinase C-epsilon and phosphodiesterase 4D3. [provided by RefSeq]

Sequence: EKTDSFYHSSGGLELYGEPRHTTYRSRSDLDYIRSPLPFQNRYPGTPADFNPGSLACSQLQNYDPDRALTDYITRLEALQRRLGTIQSGSTTQFHAGMRR
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EKTDSFYHSSGGLELYGEPRHTTYRSRSDLDYIRSPLPFQNRYPGTPADFNPGSLACSQLQNYDPDRALTDYITRLEALQRRLGTIQSGSTTQFHAGMRR
RUO
AKAP9
Wheat Germ (in vitro)
GST
NP_671700
Liquid
10142
AKAP9 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AKAP350/AKAP450/CG-NAP/HYPERION/KIAA0803/MU-RMS-40.16A/PRKA9/YOTIAO
AKAP9
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit