Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ALDH1L1 Partial ORF (NP_036322, 803 a.a. - 902 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Supplier:  Abnova™ H00010840Q01S

 View more versions of this product

Catalog No. 89-933-097


Add to Cart

Description

Description

The protein encoded by this gene catalyzes the conversion of 10-formyltetrahydrofolate, NADP, and water to tetrahydrofolate, NADPH, and carbon dioxide. The encoded protein belongs to the aldehyde dehydrogenase family and is responsible for formate oxidation in vivo. Deficiencies in this gene can result in an accumulation of formate and subsequent methanol poisoning. [provided by RefSeq]

Sequence: EESFGPVMIISRFADGDLDAVLSRANATEFGLASGVFTRDINKALYVSDKLQAGTVFVNTYNKTDVAAPFGGFKQSGFGKDLGEAALNEYLRVKTVTFEY
Specifications

Specifications

NP_036322
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EESFGPVMIISRFADGDLDAVLSRANATEFGLASGVFTRDINKALYVSDKLQAGTVFVNTYNKTDVAAPFGGFKQSGFGKDLGEAALNEYLRVKTVTFEY
RUO
ALDH1L1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
10840
ALDH1L1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp781N0997/FTHFD
ALDH1L1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit