Learn More
Abnova™ Human ALDH3A1 (AAH04370, 1 a.a. - 453 a.a.) Full-length Recombinant Protein with His tag
Used for Func, SDS-PAGE
Supplier: Abnova™ P3496
Description
Aldehyde dehydrogenases oxidize various aldehydes to the corresponding acids. They are involved in the detoxification of alcohol-derived acetaldehyde and in the metabolism of corticosteroids, biogenic amines, neurotransmitters, and lipid peroxidation. The enzyme encoded by this gene forms a cytoplasmic homodimer that preferentially oxidizes aromatic and medium-chain (6 carbons or more) saturated and unsaturated aldehyde substrates. It is thought to promote resistance to UV and 4-hydroxy-2-nonenal-induced oxidative damage in the cornea. The gene is located within the Smith-Magenis syndrome region on chromosome 17. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq]
Sequence: MGSSHHHHHHSSGLVPRGSHMSKISEAVKRARAAFSSGRTRPLQFRIQQLEALQRLIQEQEQELVGALAADLHKNEWNAYYEEVVYVLEEIEYMIQKLPEWAADEPVEKTPQTQQDELYIHSEPLGVVLVIGTWNYPFNLTIQPMVGAIAAGNAVVLKPSELSENMASLLATIIPQYLDKDLYPVINGGVPETTELLKERFDHILYTGSTGVGKIIMTAAAKHLTPVTLELGGKSPCYVDKNCDLDVACRRIAWGKFMNSGQTCVAPDYILCDPSIQNQIVEKLKKSLKEFYGEDAKKSRDYGRIISARHFQRVMGLIEGQKVAYGGTGDAATRYIAPTILTDVDPQSPVMQEEIFGPVLPIVCVRSLEEAIQFINQREKPLALYMFSSNDKVIKKMIAETSSGGVAANDVIVHITLHSLPFGGVGNSGMGSYHGKKSFETFSHRRSCLVRPLMNDEGLKVRYPPSPAKMTQHSpecifications
AAH04370 | |
Functional Study, SDS-PAGE | |
218 | |
ALDH3A1 (Human) Recombinant Protein | |
Conventional Chromatography | |
50 ug | |
RUO | |
ALDH3A1 | |
Specific activity is < 1 units/mL and was obtained by measuring the increase of NADP in absorbance at 340nm resulting from the reduction of NAD. One unit will oxidize 1.0μmole of acetaldehyde to acetic acid per minute at pH 8.0 at 25°C in the pre | |
Recombinant | |
Escherichia coli expression system | |
>95% by SDS-PAGE |
0.5 mg/mL | |
In 20mM Tris-HCl, 0.1M NaCl, pH 8.0 (10% glycerol) | |
52.5kDa | |
Escherichia coli expression system | |
Loading 3 ug protein in 15% SDS-PAGE | |
Store at 4°C for 1-2 weeks. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. | |
ALDH3/ALDHIII/MGC10406 | |
ALDH3A1 | |
E. coli | |
His | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.