Learn More
Abnova™ Human ALF Full-length ORF (NP_006863.2, 1 a.a. - 478 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_006863.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 11036 |
Molecular Weight (g/mol) | 78.8kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-933-144
|
Abnova™
H00011036P01L |
25 ug |
Each for $769.00
|
|
|||||
89-933-143
|
Abnova™
H00011036P01S |
10 ug |
Each for $499.00
|
|
|||||
Description
The assembly and stability of the RNA polymerase II transcription pre-initiation complex on a eukaryotic core promoter involve the effects of TFIIA on the interaction between TATA-binding protein (TBP) and DNA. This gene encodes a germ cell-specific counterpart of the large (alpha/beta) subunit of general transcription factor TFIIA that is able to stabilize the binding of TBP to DNA and may be uniquely important to testis biology. Alternative splicing for this locus has been observed and two variants, encoding distinct isoforms, have been identified. Co-transcription of this gene and the neighboring upstream gene generates a rare transcript (SALF), which encodes a fusion protein comprised of sequence sharing identity with each individual gene product. [provided by RefSeq]
Sequence: MACLNPVPKLYRSVIEDVIEGVRNLFAEEGIEEQVLKDLKQLWETKVLQSKATEDFFRNSIQSPLFTLQLPHSLHQTLQSSTASLVIPAGRTLPSFTTAELGTSNSSANFTFPGYPIHVPAGVTLQTVSGHLYKVNVPIMVTETSGRAGILQHPIQQVFQQLGQPSVIQTSVPQLNPWSLQATTEKSQRIETVLQQPAILPSGPVDRKHLENATSDILVSPGNEHKIVPEALLCHQESSHYISLPGVVFSPQVSQTNSNVESVLSGSASMAQNLHDESLSTSPHGALHQHVTDIQLHILKNRMYGCDSVKQPRNIEEPSNIPVSEKDSNSQVDLSIRVTDDDIGEIIQVDGSGDTSSNEEIGSTRDADENEFLGNIDGGDLKVPEEEADSISNEDSATNSSDNEDPQVNIVEEDPLNSGDDVSEQDVPDLFDTDNVIVCQYDKIHRSKNKWKFYLKDGVMCFGGRDYVFAKAIGDAEWSpecifications
NP_006863.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
78.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
MACLNPVPKLYRSVIEDVIEGVRNLFAEEGIEEQVLKDLKQLWETKVLQSKATEDFFRNSIQSPLFTLQLPHSLHQTLQSSTASLVIPAGRTLPSFTTAELGTSNSSANFTFPGYPIHVPAGVTLQTVSGHLYKVNVPIMVTETSGRAGILQHPIQQVFQQLGQPSVIQTSVPQLNPWSLQATTEKSQRIETVLQQPAILPSGPVDRKHLENATSDILVSPGNEHKIVPEALLCHQESSHYISLPGVVFSPQVSQTNSNVESVLSGSASMAQNLHDESLSTSPHGALHQHVTDIQLHILKNRMYGCDSVKQPRNIEEPSNIPVSEKDSNSQVDLSIRVTDDDIGEIIQVDGSGDTSSNEEIGSTRDADENEFLGNIDGGDLKVPEEEADSISNEDSATNSSDNEDPQVNIVEEDPLNSGDDVSEQDVPDLFDTDNVIVCQYDKIHRSKNKWKFYLKDGVMCFGGRDYVFAKAIGDAEW | |
RUO | |
GTF2A1L | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
11036 | |
ALF (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ALF/MGC26254 | |
GTF2A1L | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.