Learn More
Abnova™ Human ALG8 Partial ORF (NP_076984, 260 a.a. - 334 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00079053Q01L
Description
This gene encodes a member of the ALG6/ALG8 glucosyltransferase family. The encoded protein catalyzes the addition of the second glucose residue to the lipid-linked oligosaccharide precursor for N-linked glycosylation of proteins. Mutations in this gene have been associated with congenital disorder of glycosylation type Ih (CDG-Ih). Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: NQLPQVFSRLFPFKRGLCHAYWAPNFWALYNALDKVLSVIGLKLKFLDPNNIPKASMTSGLVQQFQHTVLPSVTPSpecifications
NP_076984 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.99kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NQLPQVFSRLFPFKRGLCHAYWAPNFWALYNALDKVLSVIGLKLKFLDPNNIPKASMTSGLVQQFQHTVLPSVTP | |
RUO | |
ALG8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
79053 | |
ALG8 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC2840 | |
ALG8 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.