Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ALK (BAG10812.1, 1058 a.a. - 1620 a.a.) F1174L mutant Partial Recombinant Protein with GST-tag expressed in Baculovirus infected Sf21 cells

Used for Func, SDS-PAGE

Manufacturer:  Abnova™ P5741

Catalog No. 89-933-174


Add to cart

Description

Description

The 2;5 chromosomal translocation is frequently associated with anaplastic large cell lymphomas (ALCLs). The translocation creates a fusion gene consisting of the ALK (anaplastic lymphoma kinase) gene and the nucleophosmin (NPM) gene: the 3' half of ALK, derived from chromosome 2, is fused to the 5' portion of NPM from chromosome 5. A recent study shows that the product of the NPM-ALK fusion gene is oncogenic. The deduced amino acid sequences reveal that ALK is a novel receptor protein-tyrosine kinase having a putative transmembrane domain and an extracellular domain. These sequences are absent in the product of the transforming NPM-ALK gene. ALK shows the greatest sequence similarity to LTK (leukocyte tyrosine kinase). ALK plays an important role in the development of the brain and exerts its effects on specific neurons in the nervous system. [provided by RefSeq]

Specifications

Specifications

Baculovirus infected insect cell (Sf21) expression system
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
90kDa
Baculovirus infected insect cell (Sf21) expression system
58% by SDS-PAGE/CBB staining
5 ug
RUO
ALK
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg (or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip™3000.
Substrate : Srctide. ATP: 100 µM.
Yes
BAG10812.1
Liquid
238
ALK (F1174L) (Human) Recombinant Protein
Glutathione sepharose chromatography
SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD246/Ki-1/TFG/ALK
ALK
Insect
GST
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit