Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ALOXE3 Full-length ORF (AAI01939.1, 1 a.a. - 711 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00059344P01S

 View more versions of this product

Catalog No. 89-933-226


Add to cart

Description

Description

This gene is a member of the lipoxygenase family, which are catabolized by arachidonic acid-derived compounds. The encoded enzyme is a hydroperoxide isomerase that synthesizes a unique type of epoxy alcohol (8R-hydroxy-11R,12R-epoxyeicosa-5Z,9E,14Z-trienoic acid) from 12R-hydroperoxyeicosatetraenoic acid (12R-HPETE). This epoxy alcohol can activate the the nuclear receptor peroxisome proliferator-activated receptor alpha (PPARalpha), which is implicated in epidermal differentiation. Loss of function of the enzyme encoded by this gene results in ichthyosis, implicating the function of this gene in the differentiation of human skin. This gene is part of a cluster of lipoxygenase genes on 17p13.1. Mutations in this gene result in nonbullous congenital ichthyosiform erythroderma (NCIE). Multiple transcript variants encoding different isoforms have been found for this gene

Sequence: MAVYRLCVTTGPYLRAGTLDNISVTLVGTCGESPKQRLDRMGRDFAPGSVQKYKVRCTAELGELLLLRVHKERYAFFRKDSWYCSRICVTEPDGSVSHFPCYQWIEGYCTVELRPGTARTICQDSLPLLLDHRTRELRARQECYRWKIYAPGFPCMVDVNSFQEMESDKKFALTKTTTCVDQGDSSGNRYLPGFPMKIDIPSLMYMEPNVRYSATKTISLLFNAIPASLGMKLRGLLDRKGSWKKLDDMQNIFWCHKTFTTKYVTEHWCEDHFFGYQYLNGVNPVMLHCISSLPSKLPVTNDMVAPLLGQDTCLQTELERGNIFLADYWILAEAPTHCLNGRQQYVAAPLCLLWLSPQGALVPLAIQLSQTPGPDSPIFLPTDSEWDWLLAKTWVRNSEFLVHENNTHFLCTHLLCEAFAMATLRQLPLCHPIYKLLLPHTRYTLQVNTIARATLLNPEGLVDQVTSIGRQGLIYLMSTGLAHFTYTNFCLPDSLRARGVLAIPNYHYRDDGLKIWAAIESFVSEIVGYYYPSDASVQQDSELQAWTGEIFAQAFLGRESSGFPSRLCTPGEMVKFLTAIIFNCSAQHAAVNSGQHDFGAWMPNAPSSMRQPPPQTKGTTTLKTYLDTLPEVNISCNNLLLFWLVSQEPKDQRPLGTYPDEHFTEEAPRRSIAAFQSRLAQISRDIQERNQGLALPYTYLDPPLIENSVSI
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
106.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
ALOXE3
Wheat Germ (in vitro)
GST
AAI01939.1
Liquid
59344
ALOXE3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
E-LOX/MGC119694/MGC119695/MGC119696/eLOX3
ALOXE3
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit