Learn More
BioworldSupplier Diversity Partner Human Alpha-1 Microglobulin Protein, 20 μg DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220602421
Human Alpha-1 Microglobulin Protein (A1M) Source: Human Background : Alpha 1-microglobulin (A1M) is an immunomodulatory protein, extracted from urine of patients with chronic renal tubular proteinuria ,with a broad spectrum of possible clinical applications and seems a promising marker for evaluation of tubular function. Physical Appearance: Sterile Filtered White lyophilized (freeze-dried) powder. Formulation: Lyophilized from 0.02M NH4HCO3. Stability: Human A1M although stable at room temperature for 3 weeks, should be stored between 2-80C. Please avoid freeze-thaw cycles. Purity: Greater than 96.0%. Reconstitution: Use phosphate buffer, pH>7.0 containing 0.15M NaCl, is recommended. Usage: BioWORLD's products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or household chemicals.
Catalog No. 50-253-342