Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Human alpha-Actinin 1 Recombinant Protein
Highly purified. Generating reliable and reproducible results.
Supplier: Novus Biologicals™ NBP2597250.5MG
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-249 of Human alpha-Actinin 1 The Recombinant Human alpha-Actinin 1 Protein is derived from E. coli. The Recombinant Human alpha-Actinin 1 Protein has been validated for the following applications: SDS-Page.Specifications
1mg/mL | |
Phosphate Buffered Saline (pH 7.4) containing 10% glycerol | |
ACTN1 Recombinant Protein | |
E.coli | |
RUO | |
ACTN1 | |
Unconjugated | |
Recombinant |
PAGE | |
31.4 kDa | |
0.5mg | |
Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, −20 to −70°C as supplied. 3 months, −20 to −70°C under sterile conditions after opening. | |
actinin 1 smooth muscle, actinin, alpha 1, ACTN1 | |
Recombinant Protein | |
Human | |
>95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?
Provide Content Correction