Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human alpha-Actinin 1 Recombinant Protein

Highly purified. Generating reliable and reproducible results.

$280.50 - $783.00

Specifications

Product Type Recombinant Protein
Recombinant Yes
Regulatory Status RUO
Storage Requirements Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, −20 to −70°C as supplied. 3 months, −20 to −70°C under sterile conditions after opening.
Gene Symbol ACTN1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP972501MG View Documents Novus Biologicals™
NBP2597250.1MG
0.1mg Each for $280.50
Add to cart
 
NBP972505MG View Documents Novus Biologicals™
NBP2597250.5MG
0.5mg Each for $783.00
Add to cart
 
Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-249 of Human alpha-Actinin 1 The Recombinant Human alpha-Actinin 1 Protein is derived from E. coli. The Recombinant Human alpha-Actinin 1 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

Recombinant Protein
RUO
ACTN1
ACTN1 Recombinant Protein
Human
PAGE
31.4 kDa
Yes
Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, −20 to −70°C as supplied. 3 months, −20 to −70°C under sterile conditions after opening.
actinin 1 smooth muscle, actinin, alpha 1, ACTN1
Phosphate Buffered Saline (pH 7.4) containing 10% glycerol
E.coli
Unlabeled

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit