Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Angiopoietin-1 Quantikine ELISA Kit

Catalog No. p-4753516
Click to view available options
Quantity:
1 Plate
6 Plates
This item is not returnable. View return policy
This item is not returnable. View return policy

Human Angiopoietin-1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Angiopoietin-1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Angiopoietin-1 in cell culture supernates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), platelet-poor citrate plasma (10 ul), saliva (25 ul), human milk (25 ul).

Specifications

Assay Range 62.50 - 4000pg/mL
Assay Sensitivity 10.3pg/mL
Conjugate HRP
Cross Reactivity Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Human Milk, Plasma (EDTA, Heparin), Platelet-poor Plasma (Citrate), Saliva, Serum
Specificity Natural and Recombinant Human Angiopoietin-1
Sufficient For Sufficient materials to run ELISAs on at least one 96-well plate
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 80381
Quantity 1 Plate
Sample Volume Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Platelet-poor Citrate Plasma (10uL),Saliva (25uL),Human Milk (25uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Target B7-H3/CD276
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.