Learn More
Novus Biologicals™ Human Annexin V ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP262136
Catalog No. NBP262136
Add to cart
Description
Description
The Human Annexin V ELISA Kit (Colorimetric) measures human Annexin V in cell culture supernatants, serum and other body fluids.
Specifications
Specifications
1 Kit
6.25 to 400ng/ml
308
1.18ng/ml
Annexin V
ANXA5
Body Fluids, Cell Culture Supernatant, Serum
ELISA
ELISA Kit (Colorimetric)
Store at 4°C
Human