Learn More
Novus Biologicals™ Human Annexin V (Vac- alpha) ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP191251
Catalog No. NBP191251
Add to cart
Description
Description
The Human Annexin V ELISA Kit (Colorimetric) measures human Annexin V in cell culture supernatant, serum.
Specifications
Specifications
6.3 - 400ng/mL
308
ELISA Kit
1.2ng/mL
Human (Hu)
Annexin V
ANXA5
ELISA
Aluminium pouch(es) with a Microwell Plate coated with recombinant Annexin V, HRP-Conjugate recombinant Annexin V, Human anti-Annexin V Standard lyophilized, 800ng/mL upon reconstitution, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum
Storage of components varies. See protocol for specific instructions.
Apoptosis
Anchorin CII, annexin A5, Annexin V, annexin-5, ANX5, Calphobindin I, Endonexin II, ENX2CBP-I, Lipocortin V, PAP-I, Placental anticoagulant protein 4, Placental anticoagulant protein I, PP4, Thromboplastin inhibitor, VAC-alpha, Vascular anticoagulant-alph