Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clostridium botulinum Toxin B, Human, Monoclonal Antibody, Abnova™

human monoclonal antibody raised against full-length recombinant Clostridium Botulinum Toxin B.

Manufacturer:  Abnova Corporation MAB5415

Catalog No. 89-135-965

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

Clostridium Botulinum Toxin B
Unconjugated
ELISA (1:1000-1:2000) Western Blot (1-5 ug/mL) Dot Blot (1-5 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
Recombinant myc and His fusion protein corresponding to Clostridium Botulinum Toxin B.
RUO
Primary
This antibody specifically detects Toxin B as determined from testing bacterial samples in Dot Blot and ELISA assays.
Dot Blot, ELISA, Western Blot
Recombinant antibody raised against full length recombinant Clostridium Botulinum Toxin B.
In PBS
Human
100 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Bacteria
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only