Learn More
Aniara Diagnostica LLCSupplier Diversity Partner HUMAN ANTITHROMIN 1.5MG DFS Item
Manufacturer:  Aniara Diagnostica LLCSupplier Diversity Partner APP004B
This item MFG by HYPHEN BioMed and Dis. by Aniara Diagnostics. Highly purified human Antithrombin III (MW=58,000 Da) from human plasma, lyophilized with glycine, HEPES and sodium chloride. Specific activity > 6 IU/mg. For in vitro use only. 1 vial x 1.5 mg.
Catalog No. 50-100-9012