Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ANXA4 Partial ORF (NP_001144, 224 a.a. - 319 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00000307Q01S

 View more versions of this product

Catalog No. 89-935-156


Add to cart

Description

Description

Annexin IV (ANX4) belongs to the annexin family of calcium-dependent phospholipid binding proteins. Although their functions are still not clearly defined, several members of the annexin family have been implicated in membrane-related events along exocytotic and endocytotic pathways. ANX4 has 45 to 59% identity with other members of its family and shares a similar size and exon-intron organization. Isolated from human placenta, ANX4 encodes a protein that has possible interactions with ATP, and has in vitro anticoagulant activity and also inhibits phospholipase A2 activity. ANX4 is almost exclusively expressed in epithelial cells. [provided by RefSeq]

Sequence: QSIKSETSGSFEDALLAIVKCMRNKSAYFAEKLYKSMKGLGTDDNTLIRVMVSRAEIDMLDIRAHFKRLYGKSLYSFIKGDTSGDYRKVLLVLCGG
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.3kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QSIKSETSGSFEDALLAIVKCMRNKSAYFAEKLYKSMKGLGTDDNTLIRVMVSRAEIDMLDIRAHFKRLYGKSLYSFIKGDTSGDYRKVLLVLCGG
RUO
ANXA4
Wheat Germ (in vitro)
GST
NP_001144
Liquid
307
ANXA4 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ANX4/DKFZp686H02120/MGC75105/PIG28/ZAP36
ANXA4
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit