Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human AP1GBP1 Partial ORF (NP_009178.3, 456 a.a. - 555 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00011276Q01L
Description
This gene encodes a protein that interacts with the gamma subunit of AP1 clathrin-adaptor complex. The AP1 complex is located at the trans-Golgi network and associates specific proteins with clathrin-coated vesicles. This encoded protein may act to connect the AP1 complex to other proteins. Alternatively spliced transcript variants that encode different isoforms have been described for this gene. [provided by RefSeq]
Sequence: DDFQDFQDASKSGSLDDSFSDFQELPASSKTSNSQHGNSAPSLLMPLPGTKALPSMDKYAVFKGIAADKSSENTVPPGDPGDKYSAFRELEQTAENKPLGSpecifications
NP_009178.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DDFQDFQDASKSGSLDDSFSDFQELPASSKTSNSQHGNSAPSLLMPLPGTKALPSMDKYAVFKGIAADKSSENTVPPGDPGDKYSAFRELEQTAENKPLG | |
RUO | |
AP1GBP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
11276 | |
AP1GBP1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC104959/SYNG | |
AP1GBP1 | |
Recombinant | |
wheat germ expression system |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction