Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human AP3M1 Full-length ORF (NP_036227.1, 1 a.a. - 418 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00026985P01L

 View more versions of this product

Catalog No. 89-935-218



Description

Description

The protein encoded by this gene is the medium subunit of AP-3, which is an adaptor-related protein complex associated with the Golgi region as well as more peripheral intracellular structures. AP-3 facilitates the budding of vesicles from the Golgi membrane and may be directly involved in protein sorting to the endosomal/lysosomal system. AP-3 is a heterotetrameric protein complex composed of two large subunits (delta and beta3), a medium subunit (mu3), and a small subunit (sigma 3). Mutations in one of the large subunits of AP-3 have been associated with the Hermansky-Pudlak syndrome, a genetic disorder characterized by defective lysosome-related organelles. Alternatively spliced transcript variants encoding the same protein have been observed. [provided by RefSeq]

Sequence: MIHSLFLINCSGDIFLEKHWKSVVSQSVCDYFFEAQEKAADVENVPPVISTPHHYLISIYRDKLFFVSVIQTEVPPLFVIEFLHRVADTFQDYFGECSEAAIKDNVVIVYELLEEMLDNGFPLATESNILKELIKPPTILRSVVNSITGSSNVGDTLPTGQLSNIPWRRAGVKYTNNEAYFDVVEEIDAIIDKSGSTVFAEIQGVIDACIKLSGMPDLSLSFMNPRLLDDVSFHPCIRFKRWESERVLSFIPPDGNFRLISYRVSSQNLVAIPVYVKHSISFKENSSCGRFDITIGPKQNMGKTIEGITVTVHMPKVVLNMNLTPTQGSYTFDPVTKVLTWDVGKITPQKLPSLKGLVNLQSGAPKPEENPSLNIQFKIQQLAISGLKVNRLDMYGEKYKPFKGVKYVTKAGKFQVRT
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
73.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
AP3M1
Wheat Germ (in vitro)
GST
NP_036227.1
Liquid
26985
AP3M1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC22164
AP3M1
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit