Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human AP3M1 Full-length ORF (NP_036227.1, 1 a.a. - 418 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name AP3M1 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 73.3kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-935-218
View Documents
Abnova™
H00026985P01L
25 ug
Each for $709.50
Add to cart
 
89-935-217
View Documents
Abnova™
H00026985P01S
10 ug
Each for $460.50
Add to cart
 
Description

Description

The protein encoded by this gene is the medium subunit of AP-3, which is an adaptor-related protein complex associated with the Golgi region as well as more peripheral intracellular structures. AP-3 facilitates the budding of vesicles from the Golgi membrane and may be directly involved in protein sorting to the endosomal/lysosomal system. AP-3 is a heterotetrameric protein complex composed of two large subunits (delta and beta3), a medium subunit (mu3), and a small subunit (sigma 3). Mutations in one of the large subunits of AP-3 have been associated with the Hermansky-Pudlak syndrome, a genetic disorder characterized by defective lysosome-related organelles. Alternatively spliced transcript variants encoding the same protein have been observed. [provided by RefSeq]

Sequence: MIHSLFLINCSGDIFLEKHWKSVVSQSVCDYFFEAQEKAADVENVPPVISTPHHYLISIYRDKLFFVSVIQTEVPPLFVIEFLHRVADTFQDYFGECSEAAIKDNVVIVYELLEEMLDNGFPLATESNILKELIKPPTILRSVVNSITGSSNVGDTLPTGQLSNIPWRRAGVKYTNNEAYFDVVEEIDAIIDKSGSTVFAEIQGVIDACIKLSGMPDLSLSFMNPRLLDDVSFHPCIRFKRWESERVLSFIPPDGNFRLISYRVSSQNLVAIPVYVKHSISFKENSSCGRFDITIGPKQNMGKTIEGITVTVHMPKVVLNMNLTPTQGSYTFDPVTKVLTWDVGKITPQKLPSLKGLVNLQSGAPKPEENPSLNIQFKIQQLAISGLKVNRLDMYGEKYKPFKGVKYVTKAGKFQVRT
Specifications

Specifications

AP3M1 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
73.3kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit