Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human AP3M2 Full-length ORF (NP_006794.1, 1 a.a. - 418 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00010947P01S
Description
This gene encodes a subunit of the heterotetrameric adaptor-related protein comlex 3 (AP-3), which belongs to the adaptor complexes medium subunits family. The AP-3 complex plays a role in protein trafficking to lysosomes and specialized organelles. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq]
Sequence: MIHSLFLINSSGDIFLEKHWKSVVSRSVCDYFFEAQERATEAENVPPVIPTPHHYLLSVYRHKIFFVAVIQTEVPPLFVIEFLHRVVDTFQDYFGVCSEPVIKDNVVVVYEVLEEMLDNGFPLATESNILKELIKPPTILRTVVNTITGSTNVGDQLPTGQLSVVPWRRTGVKYTNNEAYFDVIEEIDAIIDKSGSTITAEIQGVIDACVKLTGMPDLTLSFMNPRLLDDVSFHPCVRFKRWESERILSFIPPDGNFRLLSYHVSAQNLVAIPVYVKHNISFRDSSSLGRFEITVGPKQTMGKTIEGVTVTSQMPKGVLNMSLTPSQGTHTFDPVTKMLSWDVGKINPQKLPSLKGTMSLQAGASKPDENPTINLQFKIQQLAISGLKVNRLDMYGEKYKPFKGIKYMTKAGKFQVRTSpecifications
NP_006794.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
73.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
AP3M2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10947 | |
AP3M2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AP47B/CLA20/P47B | |
AP3M2 | |
Recombinant | |
wheat germ expression system |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction