Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human APITD1 Full-length ORF (NP_954988.1, 1 a.a. - 138 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00378708P01L

 View more versions of this product

Catalog No. 89-935-284


Add to cart

Description

Description

This gene was identified in the neuroblastoma tumour suppressor candidate region on chromosome 1p36. It contains a TFIID-31 domain, similar to that found in TATA box-binding protein-associated factor, TAF(II)31, which is required for p53-mediated transcription activation. This gene was expressed at very low levels in neuroblastoma tumours, and was shown to reduce cell growth in neuroblastoma cells, suggesting that it may have a role in a cell death pathway. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq]

Sequence: MEEEAETEEQQRFSYQQRLKAAVHYTVGCLCEEVALDKEMQFSKQTIAAISELTFRQCENFAKDLEMFARHAKRTTINTEDVKLLARRSNSLLKYITDKSEEIAQINLERKAQKKKKSEDGSKNSRQPAEAGVVESEN
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
42.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
APITD1
Wheat Germ (in vitro)
GST
NP_954988.1
Liquid
378708
APITD1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CENP-S/CENPS/MGC32686
APITD1
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit