Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human APOBEC3B Full-length ORF (AAH53859.1, 1 a.a. - 251 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00009582P01L

 View more versions of this product

Catalog No. 89-935-319


Add to cart

Description

Description

This gene is a member of the cytidine deaminase gene family. It is one of seven related genes or pseudogenes found in a cluster, thought to result from gene duplication, on chromosome 22. Members of the cluster encode proteins that are structurally and functionally related to the C to U RNA-editing cytidine deaminase APOBEC1. It is thought that the proteins may be RNA editing enzymes and have roles in growth or cell cycle control. [provided by RefSeq]

Sequence: MNPQIRNPMERMYRDTFYDNFENEPILYGRSYTWLCYEVKIKRGRSNLLWDTGVFRGQVYFEPQYHAEMCFLSWFCGNQLPAYKCFQITWFVSWTPCPDCVAKLAEFLSEHPNVTLTISAARLYYYWERDYRRALCRLSQAGARVKIMDYEEFAYCWENFVYNEGQQFMPWYKFDENYAFLHRTLKEILRLRIFSVAFTAAMRSCASWTWFLLCSWTRPRSTGSLGSSPGAPASPGAVPGKCVRSFRRTHT
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
56.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
APOBEC3B
Wheat Germ (in vitro)
GST
AAH53859.1
Liquid
9582
APOBEC3B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
APOBEC1L/ARCD3/ARP4/DJ742C19.2/FLJ21201/PHRBNL/bK150C2.2
APOBEC3B
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit