Learn More
Abnova™ Human APOD (P05089) Partial Recombinant Protein
Used for SDS-PAGE
Manufacturer:  Abnova™ P4052
Catalog No. 89-935-353
Add to cart
Description
Description
This gene encodes a component of high density lipoprotein that has no marked similarity to other apolipoprotein sequences. It has a high degree of homology to plasma retinol-binding protein and other members of the alpha 2 microglobulin protein superfamily of carrier proteins, also known as lipocalins. This glycoprotein is closely associated with the enzyme lecithin:cholesterol acyltransferase - an enzyme involved in lipoprotein metabolism. [provided by RefSeq]
Specifications
Specifications
Escherichia coli expression system
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
19.8kDa
Escherichia coli expression system
10 ug
RUO
APOD
Escherichia coli
None
P05089
Lyophilized
347
APOD (Human) Recombinant Protein
≥95% by SDS-PAGE and HPLC
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
<0.1ng/μg
APOD
Yes