Learn More
Abnova™ Human APPBP2 Full-length ORF (NP_006371.2, 1 a.a. - 585 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00010513P01L
Description
The protein encoded by this gene interacts with microtubules and is functionally associated with beta-amyloid precursor protein transport and/or processing. The beta-amyloid precursor protein is a cell surface protein with signal-transducing properties, and it is thought to play a role in the pathogenesis of Alzheimer's disease. This gene has been found to be highly expressed in breast cancer. Multiple polyadenylation sites have been found for this gene. [provided by RefSeq]
Sequence: MAAVELEWIPETLYNTAISAVVDNYIRSRRDIRSLPENIQFDVYYKLYQQGRLCQLGSEFCELEVFAKVLRALDKRHLLHHCFQALMDHGVKVASVLAYSFSRRCSYIAESDAAVKEKAIQVGFVLGGFLSDAGWYSDAEKVFLSCLQLCTLHDEMLHWFRAVECCVRLLHVRNGNCKYHLGEETFKLAQTYMDKLSKHGQQANKAALYGELCALLFAKSHYDEAYKWCIEAMKEITAGLPVKVVVDVLRQASKACVVKREFKKAEQLIKHAVYLARDHFGSKHPKYSDTLLDYGFYLLNVDNICQSVAIYQAALDIRQSVFGGKNIHVATAHEDLAYSSYVHQYSSGKFDNALFHAERAIGIITHILPEDHLLLASSKRVKALILEEIAIDCHNKETEQRLLQEAHDLHLSSLQLAKKAFGEFNVQTAKHYGNLGRLYQSMRKFKEAEEMHIKAIQIKEQLLGQEDYEVALSVGHLASLYNYDMNQYENAEKLYLRSIAIGKKLFGEGYSGLEYDYRGLIKLYNSIGNYEKVFEYHNVLSNWNRLRDRQYSVTDALEDVSTSPQSTEEVVQSFLISQNVEGPSCSpecifications
NP_006371.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
93.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
APPBP2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10513 | |
APPBP2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HS.84084/KIAA0228/PAT1 | |
APPBP2 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.