Learn More
Abnova™ Human ARFGAP3 Full-length ORF (NP_055385.2, 1 a.a. - 516 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00026286P01S
Description
The protein encoded by this gene is a GTPase-activating protein (GAP) that associates with the Golgi apparatus and regulates the early secretory pathway of proteins. The encoded protein promotes hydrolysis of ADP-ribosylation factor 1 (ARF1)-bound GTP, which is required for the dissociation of coat proteins from Golgi-derived membranes and vesicles. Dissociation of the coat proteins is a prerequisite for the fusion of these vesicles with target compartments. The activity of this protein is sensitive to phospholipids. Multiple transcript variants encoding different isoforms have been found for this gene. This gene was originally known as ARFGAP1, but that is now the name of a related but different gene. [provided by RefSeq]
Sequence: MGDPSKQDILTIFKRLRSVPTNKVCFDCGAKNPSWASITYGVFLCIDCSGSHRSLGVHLSFIRSTELDSNWSWFQLRCMQVGGNASASSFFHQHGCSTNDTNAKYNSRAAQLYREKIKSLASQATRKHGTDLWLDSCVVPPLSPPPKEEDFFASHVSPEVSDTAWASAIAEPSSLTSRPVETTLENNEGGQEQGPSVEGLNVPTKATLEVSSIIKKKPNQAKKGLGAKKGSLGAQKLANTCFNEIEKQAQAADKMKEQEDLAKVVSKEESIVSSLRLAYKDLEIQMKKDEKMNISGKKNVDSDRLGMGFGNCRSVISHSVTSDMQTIEQESPIMAKPRKKYNDDSDDSYFTSSSRYFDEPVELRSSSFSSWDDSSDSYWKKETSKDTETVLKTTGYSDRPTARRKPDYEPVENTDEAQKKFGNVKAISSDMYFGRQSQADYETRARLERLSASSSISSADLFEEPRKQPAGNYSLSSVLPNAPDMAQFKQGVRSVAGKLSVFANGVVTSIQDRYGSSpecifications
NP_055385.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
83.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
ARFGAP3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
26286 | |
ARFGAP3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARFGAP1/FLJ45618 | |
ARFGAP3 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.