Learn More
Novus Biologicals™ Human Arg-Vasopressin ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP266644
Catalog No. NBP266644
Add to cart
Description
Description
The Human Neurophysin II/Arg-vasopressin ELISA Kit (Colorimetric) measures human Neurophysin II/Arg-vasopressin in serum, plasma and other biological fluids.
Specifications
Specifications
ELISA Kit (Colorimetric)
This kit recognizes Human ADCY4 in samples. No significant cross-reactivity or interference between Human ADCY4 and analogues was observed.
Storage is content dependent.
Human
Biological Fluids, Plasma, Serum
AVP
15.63-1000 pg/mL
ELISA
Arg-Vasopressin
1 Kit
0.38 ng/mL