Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ARHGAP28 Full-length ORF (AAH65274.1, 1 a.a. - 545 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00079822P01

Catalog No. 89-935-510


Add to cart

Description

Description

Sequence: MNELPRDTCGNHTNQLDGTKEERELPRVIKTSGSMPDDASLNSTTLSDASQDKEGSFAVPRSDSVAILETIPVLPVHSNGSPEPGQPVQNAISDDDFLEKNIPPEAEELSFEVSYSEMVTEALKRNKLKKSEIKKEDYVLTKFNVQKTRFGLTEAGDLSAEDMKKIRHLSLIELTAFFDAFGIQLKRNKTEKVKGRDNGIFGVPLTVLLDGDRKKDPGVKVPLVLQKFFEKVEESGLESEGIFRLSGCTAKVKQYREELDAKFNADKFKWDKMCHREAAVMLKAFFRELPTSLFPVEYIPAFISLMERGPHVKVQFQALHLMVMALPDANRDAAQALMTFFNKVIANESKNRMSLWNISTVMAPNLFFSRSKHSDYEELLLANTAAHIIRLMLKYQKILWKVPSFLITQVRRMNEATMLLKKQLPSVRKLLRRKTLERETASPKTSKVLQKSPSARRMSDVPEGVIRVHAPLLSKVSMAIQLNNQTKAKDILAKFQYENRILHWQRAALSFLNGKWVKKEREESTETNRSPKHVFLFTIGLDIST
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
88.4kDa
Glutathione Sepharose 4 Fast Flow
2 ug
RUO
ARHGAP28
Wheat Germ (in vitro)
GST
AAH65274.1
Liquid
79822
ARHGAP28 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686A2038/FLJ10312
ARHGAP28
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit