Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ARHGEF2 Partial ORF (NP_004714, 590 a.a. - 690 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$499.00 - $769.00

Specifications

Accession Number NP_004714
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9181
Molecular Weight (g/mol) 36.85kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-935-540
View Documents
Abnova™
H00009181Q02S
10 ug
Each for $499.00
Add to Cart
 
89-935-541
View Documents
Abnova™
H00009181Q02L
25 ug
Each for $769.00
Add to Cart
 
Description

Description

Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form complex with G proteins and stimulate rho-dependent signals. Alternatively spliced transcript variants encoding different isoforms have been identified.[provided by RefSeq]

Sequence: MTHFQAEEDGGSGMALPTLPRGLFRSESLESPRGERLLQDAIREVEGLKDLLVGPGVELLLTPREPALPLEPDSGGNTSPGVTANGEARTFNGSIELCRAD
Specifications

Specifications

NP_004714
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.85kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp547L106/DKFZp547P1516/GEF/GEF-H1/GEFH1/KIAA0651/LFP40/P40
ARHGEF2
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
9181
ARHGEF2 (Human) Recombinant Protein (Q02)
MTHFQAEEDGGSGMALPTLPRGLFRSESLESPRGERLLQDAIREVEGLKDLLVGPGVELLLTPREPALPLEPDSGGNTSPGVTANGEARTFNGSIELCRAD
RUO
ARHGEF2
Wheat Germ (in vitro)
GST
Liquid
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit