Learn More
Abnova™ Human ARHGEF3 Partial ORF (NP_062455, 33 a.a. - 142 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00050650Q01S
Description
Rho-like GTPases are involved in a variety of cellular processes, and they are activated by binding GTP and inactivated by conversion of GTP to GDP by their intrinsic GTPase activity. Guanine nucleotide exchange factors (GEFs) accelerate the GTPase activity of Rho GTPases by catalyzing their release of bound GDP. This gene encodes a guanine nucleotide exchange factor, which specifically activates two members of the Rho GTPase family: RHOA and RHOB, both of which have a role in bone cell biology. It has been identified that genetic variation in this gene plays a role in the determination of bone mineral density (BMD), indicating the implication of this gene in postmenopausal osteoporosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: EPSNKRVKPLSRVTSLANLIPPVKATPLKRFSQTLQRSISFRSESRPDILAPRPWSRNAAPSSTKRRDSKLWSETFDVCVNQMLTSKEIKRQEAIFELSQGEEDLIEDLKSpecifications
NP_062455 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EPSNKRVKPLSRVTSLANLIPPVKATPLKRFSQTLQRSISFRSESRPDILAPRPWSRNAAPSSTKRRDSKLWSETFDVCVNQMLTSKEIKRQEAIFELSQGEEDLIEDLK | |
RUO | |
ARHGEF3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
50650 | |
ARHGEF3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434F2429/FLJ98126/GEF3/MGC118905/STA3/XPLN | |
ARHGEF3 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.