Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ARHGEF9 Partial ORF (NP_056000.1, 419 a.a. - 516 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00023229Q01S

 View more versions of this product

Catalog No. 89-935-563


Add to cart

Description

Description

ARHGEF9 belongs to a family of Rho-like GTPases that act as molecular switches by cycling from the active GTP-bound state to the inactive GDP-bound state. These proteins are key regulators of the actin cytoskeleton and are involved in cell signaling.[supplied by OMIM]

Sequence: AFREERKMVQEDEKIGFEISENQKRQAAMTVRKVPKQKGVNSARSVPPSYPPPQDPLNHGQYLVPDGIAQSQVFEFTEPKRSQSPFWQNFSRLTPFKK
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AFREERKMVQEDEKIGFEISENQKRQAAMTVRKVPKQKGVNSARSVPPSYPPPQDPLNHGQYLVPDGIAQSQVFEFTEPKRSQSPFWQNFSRLTPFKK
RUO
ARHGEF9
Wheat Germ (in vitro)
GST
NP_056000.1
Liquid
23229
ARHGEF9 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
COLLYBISTIN/HPEM-2/KIAA0424/PEM-2/PEM2
ARHGEF9
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit