Learn More
Abnova™ Human ARID3B Full-length ORF (AAH60824.1, 1 a.a. - 253 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00010620P01S
Description
This gene encodes a member of the ARID (AT-rich interaction domain) family of DNA-binding proteins. The encoded protein is homologous with two proteins that bind to the retinoblastoma gene product, and also with the mouse Bright and Drosophila dead ringer proteins. A pseudogene on chromosome 1p31 exists for this gene. Members of the ARID family have roles in embryonic patterning, cell lineage gene regulation, cell cycle control, transcriptional regulation and possibly in chromatin structure modification. [provided by RefSeq]
Sequence: MEPLQQQQQQQQQQQKQPHLAPLQMDAREKQGQQMREAQFLYAQKLVTQPTLLSATAGRPSGSTPLGPLARVPPTAAVAQVFERGNMNSEPEEEDGGLEDEDGDDEVAEVAEKETQAASKYFHVQKVARQDPRVAPMSNLLPAPGLPPHGQQAKEDHTKDASKASPSVSTAGQPNWNLDEQLKQNGGLAWSDDADGGRGREISRDFAKLYELDGDPERKEFLDDLFVFMQKRGECASTHHSNSGNTDRVPTVCSpecifications
AAH60824.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
54.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
ARID3B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10620 | |
ARID3B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BDP/DRIL2 | |
ARID3B | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.