Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ARID4A Partial ORF (NP_002883, 1033 a.a. - 1139 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00005926Q01L

 View more versions of this product

Catalog No. 89-935-579


Add to cart

Description

Description

The protein encoded by this gene is a ubiquitously expressed nuclear protein. It binds directly, with several other proteins, to retinoblastoma protein (pRB) which regulates cell proliferation. pRB represses transcription by recruiting the encoded protein. This protein, in turn, serves as a bridging molecule to recruit HDACs and, in addition, provides a second HDAC-independent repression function. The encoded protein possesses transcriptional repression activity. Multiple alternatively spliced transcripts have been observed for this gene, although not all transcript variants have been fully described. [provided by RefSeq]

Sequence: AEESQEGLCERESANGFETNVASGTCSIIVQERESREKGQKRPSDGNSGLMAKKQKRTPKRTSAAAKNEKNGTGQSSDSEDLPVLDNSSKCTPVKHLNVSKPQKLAR
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.51kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AEESQEGLCERESANGFETNVASGTCSIIVQERESREKGQKRPSDGNSGLMAKKQKRTPKRTSAAAKNEKNGTGQSSDSEDLPVLDNSSKCTPVKHLNVSKPQKLAR
RUO
ARID4A
Wheat Germ (in vitro)
GST
NP_002883
Liquid
5926
ARID4A (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RBBP1/RBP-1/RBP1
ARID4A
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit