Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ARL4D Full-length ORF (NP_001652.2, 1 a.a. - 201 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00000379P01S

 View more versions of this product

Catalog No. 89-935-605


Add to cart

Description

Description

ADP-ribosylation factor 4D is a member of the ADP-ribosylation factor family of GTP-binding proteins. ARL4D is closely similar to ARL4A and ARL4C and each has a nuclear localization signal and an unusually high guanine nucleotide exchange rate. This protein may play a role in membrane-associated intracellular trafficking. Mutations in this gene have been associated with Bardet-Biedl syndrome (BBS). [provided by RefSeq]

Sequence: MGNHLTEMAPTASSFLPHFQALHVVVIGLDSAGKTSLLYRLKFKEFVQSVPTKGFNTEKIRVPLGGSRGITFQVWDVGGQEKLRPLWRSYTRRTDGLVFVVDAAEAERLEEAKVELHRISRASDNQGVPVLVLANKQDQPGALSAAEVEKRLAVRELAAATLTHVQGCSAVDGLGLQQGLERLYEMILKRKKAARGGKKRR
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
48.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
ARL4D
Wheat Germ (in vitro)
GST
NP_001652.2
Liquid
379
ARL4D (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ARF4L/ARL6
ARL4D
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit