Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ARMCX2 Partial ORF (NP_055597, 508 a.a. - 599 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name ARMCX2 (Human) Recombinant Protein (Q01)
Recombinant Yes
For Use With (Application) Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Molecular Weight 35.86kDa
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-935-650
View Documents
Abnova™
H00009823Q01L
25 ug
Each for $709.50
Add to cart
 
89-935-649
View Documents
Abnova™
H00009823Q01S
10 ug
Each for $460.50
Add to cart
 
Description

Description

This gene encodes a member of the ALEX family of proteins and may play a role in tumor suppression. The encoded protein contains a potential N-terminal transmembrane domain and a single Armadillo (arm) repeat. Other proteins containing the arm repeat are involved in development, maintenance of tissue integrity, and tumorigenesis. This gene is closely localized with other family members, including ALEX1 and ALEX3, on the X chromosome. Two alternative transcripts that encode the same protein but differ in the 5' UTR have been described. Additional alternative transcripts may exist but their full length natures have not been determined. A pseudogene for this locus is located on chromosome 7. [provided by RefSeq]

Sequence: NSIANFFRLLSQGGGKIKVEILKILSNFAENPDMLKKLLSTQVPASFSSLYNSYVESEILINALTLFEIIYDNLRAEVFNYREFNKGSLFYL
Specifications

Specifications

ARMCX2 (Human) Recombinant Protein (Q01)
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Yes
35.86kDa
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit