Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ARNT2 Full-length ORF (AAH36099, 1 a.a. - 716 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name ARNT2 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 104.28kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-935-674
View Documents
Abnova™
H00009915P01S
10 ug
Each for $460.50
Add to cart
 
89-935-675
View Documents
Abnova™
H00009915P01L
25 ug
Each for $709.50
Add to cart
 
Description

Description

This gene encodes a member of the basic-helix-loop-helix-Per-Arnt-Sim (bHLH-PAS) superfamily of transcription factors. The encoded protein acts as a partner for several sensor proteins of the bHLH-PAS family, forming heterodimers with the sensor proteins that bind regulatory DNA sequences in genes responsive to developmental and environmental stimuli. Under hypoxic conditions, the encoded protein complexes with hypoxia-inducible factor 1alpha in the nucleus and this complex binds to hypoxia-responsive elements in enhancers and promoters of oxygen-responsive genes. A highly similar protein in mouse forms functional complexes with both aryl hydrocarbon receptors and Single-minded proteins, suggesting addition roles for the encoded protein in the metabolism of xenobiotic compounds and the regulation of neurogenesis, respectively. [provided by RefSeq]

Sequence: MATPAAVNPPEMASDIPGSVTLPVAPMAATGQVRMAGAMPARGGKRRSGMDFDDEDGEGPSKFSRENHSEIERRRRNKMTQYITELSDMVPTCSALARKPDKLTILRMAVSHMKSMRGTGNKSTDGAYKPSFLTEQELKHLILEAADGFLFVVAAETGRVIYVSDSVTPVLNQPQSEWFGSTLYEQVHPDDVEKLREQLCTSENSMTGRILDLKTGTVKKEGQQSSMRMCMGSRRSFICRMRCGNAPLDHLPLNRITTMRKRFRNGLGPVKEGEAQYAVVHCTGYIKAWPPAGMTIPEEDADVGQGSKYCLVAIGRLQVTSSPVCMDMNGMSVPTEFLSRHNSDGIITFVDPRCISVIGYQPQDLLGKDILEFCHPEDQSHLRESFQQVVKLKGQVLSVMYRFRTKNREWMLIRTSSFTFQNPYSDEIEYIICTNTNVKQLQQQQAELEVHQRDGLSSYDLSQVPVPNLPAGVHEAGKSVEKADAIFSQERDPRFAEMFAGISASEKKMMSSASAAGTQQIYSQGSPFPSGHSGKAFSSSVVHVPGVNDIQSSSSTGQNMSQISRQLNQSQVAWTGSRPPFPGQQIPSQSSKTQSSPFGIGTSHTYPADPSSYSPLSSPATSSPSGNAYSSLANRTPGFAESGQSSGQFQGRPSEVWSQWQSQHHGQQSGEQHSHQQPGQTEVFQDMLPMPGDPGNEWWSPHSRQHFRQPISYARM
Specifications

Specifications

ARNT2 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
104.28kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit