Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human ARPC3 Partial ORF (NP_005710.1, 1 a.a. - 78 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00010094Q02L
Description
This gene encodes one of seven subunits of the human Arp2/3 protein complex. The Arp2/3 protein complex has been implicated in the control of actin polymerization in cells and has been conserved through evolution. The exact role of the protein encoded by this gene, the p21 subunit, has yet to be determined. [provided by RefSeq]
Sequence: MPAYHSSLMDPDTKLIGNMALLPIRSQFKGPAPRETKDTDIVDEAIYYFKANVFFKNYEIKNEADRTLIYITLYISECSpecifications
NP_005710.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.32kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPAYHSSLMDPDTKLIGNMALLPIRSQFKGPAPRETKDTDIVDEAIYYFKANVFFKNYEIKNEADRTLIYITLYISEC | |
RUO | |
ARPC3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10094 | |
ARPC3 (Human) Recombinant Protein (Q02) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARC21/p21-Arc | |
ARPC3 | |
Recombinant | |
wheat germ expression system |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction