Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ARPC5 Partial ORF (AAH57237.1, 31 a.a. - 89 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00010092Q01L

 View more versions of this product

Catalog No. 89-935-693


Add to cart

Description

Description

This gene encodes one of seven subunits of the human Arp2/3 protein complex. The Arp2/3 protein complex has been implicated in the control of actin polymerization in cells and has been conserved through evolution. The exact role of the protein encoded by this gene, the p16 subunit, has yet to be determined. [provided by RefSeq]

Sequence: GDGQAGPDEGEVDSCLRHSITGNMTAALQAALKNPPINTKSQAVKDRAGSIVLKVLISF
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.23kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GDGQAGPDEGEVDSCLRHSITGNMTAALQAALKNPPINTKSQAVKDRAGSIVLKVLISF
RUO
ARPC5
Wheat Germ (in vitro)
GST
AAH57237.1
Liquid
10092
ARPC5 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ARC16/MGC88523/dJ127C7.3/p16-Arc
ARPC5
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit