Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ARSE Partial ORF (NP_000038, 490 a.a. - 589 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00000415Q01S

 View more versions of this product

Catalog No. 89-935-741



Description

Description

Arylsulfatase E is a member of the sulfatase family. It is glycosylated postranslationally and localized to the golgi apparatus. Sulfatases are essential for the correct composition of bone and cartilage matrix. X-linked chondrodysplasia punctata, a disease characterized by abnormalities in cartilage and bone development, has been linked to mutations in this gene. [provided by RefSeq]

Sequence: GACYGRKVCPCFGEKVVHHDPPLLFDLSRDPSETHILTPASEPVFYQVMERVQQAVWEHQRTLSPVPLQLDRLGNIWRPWLQPCCGPFPLCWCLREDDPQ
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GACYGRKVCPCFGEKVVHHDPPLLFDLSRDPSETHILTPASEPVFYQVMERVQQAVWEHQRTLSPVPLQLDRLGNIWRPWLQPCCGPFPLCWCLREDDPQ
RUO
ARSE
Wheat Germ (in vitro)
GST
NP_000038
Liquid
415
ARSE (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CDPX/CDPX1/CDPXR/MGC163310
ARSE
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit