Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ARSF Partial ORF (NP_004033, 101 a.a. - 210 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00000416Q01S

 View more versions of this product

Catalog No. 89-935-743


Add to cart

Description

Description

This gene is a member of the sulfatase family, and more specifically, the arylsulfatase subfamily. Members of the subfamily share similarity in sequence and splice sites, and are clustered together on chromosome X, suggesting that they are derived from recent gene duplication events. Sulfatases are essential for the correct composition of bone and cartilage matrix. The activity of this protein, unlike that of arylsulfatase E, is not inhibited by warfarin. [provided by RefSeq]

Sequence: NRRVIQNLAVPAGLPLNETTLAALLKKQGYSTGLIGKWHQGLNCDSRSDQCHHPYNYGFDYYYGMPFTLVDSCWPDPSRNTELAFESQLWLCVQLVAIAILTLTFGKLSG
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
NRRVIQNLAVPAGLPLNETTLAALLKKQGYSTGLIGKWHQGLNCDSRSDQCHHPYNYGFDYYYGMPFTLVDSCWPDPSRNTELAFESQLWLCVQLVAIAILTLTFGKLSG
RUO
ARSF
Wheat Germ (in vitro)
GST
NP_004033
Liquid
416
ARSF (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ASF
ARSF
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit