Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human ARSH Full-length ORF (AAI53086.1, 1 a.a. - 562 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00347527P01S
Description
Sulfatases, such as ARSH, hydrolyze sulfate esters from sulfated steroids, carbohydrates, proteoglycans, and glycolipids. They are involved in hormone biosynthesis, modulation of cell signaling, and degradation of macromolecules (Sardiello et al., 2005 [PubMed 16174644]).[supplied by OMIM]
Sequence: MTRNARPNIVLLMADDLGVGDLCCYGNNSVSTPNIDRLASEGVRLTQHLAAASMCTPSRAAFLTGRYPIRSGMVSAYNLNRAFTWLGGSGGLPTNETTFAKLLQHRGYRTGLIGKWHLGLSCASRNDHCYHPLNHGFHYFYGVPFGLLSDCQASKTPELHRWLRIKLWISTVALALVPFLLLIPKFARWFSVPWKVIFVFALLAFLFFTSWYSSYGFTRRWNCILMRNHEIIQQPMKEEKVASLMLKEALAFIERYKREPFLLFFSFLHVHTPLISKKKFVGRSKYGRYGDNVEEMDWMVGKILDALDQERLANHTLVYFTSDNGGHLEPLDGAVQLGGWNGIYKGGKGMGGWEGGIRVPGIFRWPSVLEAGRVINEPTSLMDIYPTLSYIGGGILSQDRVIDGQNLMPLLEGRASHSDHEFLFHYCGVYLHTVRWHQKDCATVWKAHYVTPKFYPEGTGACYGSGICSCSGDVTYHDPPLLFDISRDPSEALPLNPDNEPLFDSVIKKMEAAIREHRRTLTPVPQQFSVFNTIWKPWLQPCCGTFPFCGCDKEDDILPMAPSpecifications
AAI53086.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
88.77kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
ARSH | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
347527 | |
ARSH (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
sulfatase | |
ARSH | |
Recombinant | |
wheat germ expression system |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction