Learn More
Abnova™ Human ASAH1 Partial ORF (NP_808592, 25 a.a. - 124 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00000427Q01L
Description
This gene encodes a heterodimeric protein consisting of a nonglycosylated alpha subunit and a glycosylated beta subunit that is cleaved to the mature enzyme posttranslationally. The encoded protein catalyzes the synthesis and degradation of ceramide into sphingosine and fatty acid. Mutations in this gene have been associated with a lysosomal storage disorder known as Farber disease. Multiple transcript variants encoding several distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: PPWTEDCRKSTYPPSGPTYRGAVPWYTINLDLPPYKRWHELMLDKAPMLKVIVNSLKNMINTFVPSGKVMQVVDEKLPGLLGNFPGPFEEEMKGIAAVTDSpecifications
NP_808592 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PPWTEDCRKSTYPPSGPTYRGAVPWYTINLDLPPYKRWHELMLDKAPMLKVIVNSLKNMINTFVPSGKVMQVVDEKLPGLLGNFPGPFEEEMKGIAAVTD | |
RUO | |
ASAH1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
427 | |
ASAH1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AC/ASAH/FLJ21558/FLJ22079/PHP/PHP32 | |
ASAH1 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.