Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ASB11 Partial ORF (NP_543149, 214 a.a. - 323 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00140456Q01S

 View more versions of this product

Catalog No. 89-935-794


Add to cart

Description

Description

The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing (ASB) family of proteins. They contain ankyrin repeat sequence and SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling (SOCS) proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: KLLELGASVDHGQWLDTPLHAAARQSNVEVIHLLTDYGANLKRRNAQGKSALDLAAPKSSVEQALLLREGPPALSQLCRLCVRKCLGRACHQAIHKLHLPEPLERFLLYQ
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KLLELGASVDHGQWLDTPLHAAARQSNVEVIHLLTDYGANLKRRNAQGKSALDLAAPKSSVEQALLLREGPPALSQLCRLCVRKCLGRACHQAIHKLHLPEPLERFLLYQ
RUO
ASB11
Wheat Germ (in vitro)
GST
NP_543149
Liquid
140456
ASB11 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp779E2460/MGC119168/MGC119169
ASB11
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit