Learn More
Abnova™ Human ASB11 Partial ORF (NP_543149, 214 a.a. - 323 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_543149 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 140456 |
Molecular Weight (g/mol) | 37.84kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-935-795
|
Abnova™
H00140456Q01L |
25 ug |
Each for $769.00
|
|
|||||
89-935-794
|
Abnova™
H00140456Q01S |
10 ug |
Each for $499.00
|
|
|||||
Description
The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing (ASB) family of proteins. They contain ankyrin repeat sequence and SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling (SOCS) proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: KLLELGASVDHGQWLDTPLHAAARQSNVEVIHLLTDYGANLKRRNAQGKSALDLAAPKSSVEQALLLREGPPALSQLCRLCVRKCLGRACHQAIHKLHLPEPLERFLLYQSpecifications
NP_543149 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp779E2460/MGC119168/MGC119169 | |
ASB11 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
140456 | |
ASB11 (Human) Recombinant Protein (Q01) | |
KLLELGASVDHGQWLDTPLHAAARQSNVEVIHLLTDYGANLKRRNAQGKSALDLAAPKSSVEQALLLREGPPALSQLCRLCVRKCLGRACHQAIHKLHLPEPLERFLLYQ | |
RUO | |
ASB11 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.