Learn More
Abnova™ Human ASB4 Partial ORF (NP_057200, 295 a.a. - 403 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00051666Q01S
Description
The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing (ASB) family of proteins. They contain ankyrin repeat sequence and SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling (SOCS) proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. Multiple alternatively spliced transcript variants have been described for this gene but some of the full length sequences are not known. [provided by RefSeq]
Sequence: VTSVRPAAQPEICYQLLLNHGAARIYPPQFHKVIQACHSCPKAIEVVVNAYEHIRWNTKWRRAIPDDDLEKYWDFYHSLFTVCCNSPRTLMHLSRCAIRRTLHNRCHRASpecifications
NP_057200 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VTSVRPAAQPEICYQLLLNHGAARIYPPQFHKVIQACHSCPKAIEVVVNAYEHIRWNTKWRRAIPDDDLEKYWDFYHSLFTVCCNSPRTLMHLSRCAIRRTLHNRCHRA | |
RUO | |
ASB4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
51666 | |
ASB4 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ASB-4/MGC142039/MGC142041 | |
ASB4 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.