Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ASCL3 Partial ORF (NP_065697.1, 1 a.a. - 95 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Supplier:  Abnova™ H00056676Q01S

 View more versions of this product

Catalog No. 89-935-830


Add to Cart

Description

Description

Basic helix-loop-helix transcription factors, such as ASCL3, are essential for the determination of cell fate and the development and differentiation of numerous tissues (Jonsson et al., 2004 [PubMed 15475265]).[supplied by OMIM]

Sequence: MMDNRGNSSLPDKLPIFPDSARLPLTRSFYLEPMVTFHVHPEAPVSSPYSEELPRLPFPSDSLILGNYSEPCPFSFPMPYPNYRGCEYSYGPAFT
Specifications

Specifications

NP_065697.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.19kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MMDNRGNSSLPDKLPIFPDSARLPLTRSFYLEPMVTFHVHPEAPVSSPYSEELPRLPFPSDSLILGNYSEPCPFSFPMPYPNYRGCEYSYGPAFT
RUO
ASCL3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
56676
ASCL3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HASH3/SGN1/bHLHa42
ASCL3
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit