Learn More
Human ATBF1/ZFHX3 Antibody, R&D Systems™
GREENER_CHOICE
Sheep Polyclonal Antibody
Supplier:  R&D Systems AF7384SP
Catalog No. AF7384SP
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
ATBF1/ZFHX3 Polyclonal specifically detects ATBF1/ZFHX3 in Human samples. It is validated for Immunocytochemistry.
Specifications
Specifications
ATBF1/ZFHX3
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Alpha-fetoprotein enhancer-binding protein, AT motif-binding factor, AT motif-binding factor 1, ATBF1alpha-fetoprotein enhancer binding protein, AT-binding transcription factor 1ATBT, ATBT, ZFH3, ZFH-3, ZFHX3, zinc finger homeobox 3, zinc finger homeobox protein 3, Zinc finger homeodomain protein 3, ZNF927
Sheep
Affinity Purified
RUO
463
Sterile PBS to a final concentration of 0.2 mg/mL.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Immunocytochemistry
LYOPH
Immunocytochemistry 5-15 ug/mL
Q15911
ZFHX3
E. coli-derived recombinant human ATBF1/ZFHX3 His3608-Leu3703 Accession # Q15911
25 μg
Primary
Detects human ATBF1/ZFHX3 in direct ELISAs.
Human
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only