Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ATCAY Partial ORF (NP_149053.1, 3 a.a. - 68 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00085300Q01S

 View more versions of this product

Catalog No. 89-935-907


Add to cart

Description

Description

This gene encodes a neuron-restricted protein that contains a CRAL-TRIO motif common to proteins that bind small lipophilic molecules. Mutations in this gene are associated with cerebellar ataxia, Cayman type. [provided by RefSeq]

Sequence: TTEATLRMENVDVKEEWQDEDLPRPLPEETGVELLGSPVEDTSSPPNTLNFNGAHRKRKTLVAPEI
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TTEATLRMENVDVKEEWQDEDLPRPLPEETGVELLGSPVEDTSSPPNTLNFNGAHRKRKTLVAPEI
RUO
ATCAY
Wheat Germ (in vitro)
GST
NP_149053.1
Liquid
85300
ATCAY (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BNIP-H/CLAC/KIAA1872
ATCAY
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit