Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human ATE1 Partial ORF (NP_001001976, 1 a.a. - 87 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00011101Q01L
Description
This gene encodes an arginyltransferase, an enzyme that is involved in posttranslational conjugation of arginine to N-terminal aspartate or glutamate residues. Conjugation of arginine to the N-terminal aspartate or glutamate targets proteins for ubiquitin-dependent degradation. Alternative splicing results in two transcript variants encoding distinct isoforms. [provided by RefSeq]
Sequence: MAFWAGGSPSVVDYFPSEDFYRCGYCKNESGSRSNGMWAHSMTVQDYQDLIDRGWRRSGKYVYKPVMNQTCCPQYTIRCRPLQFQPSSpecifications
NP_001001976 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.31kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAFWAGGSPSVVDYFPSEDFYRCGYCKNESGSRSNGMWAHSMTVQDYQDLIDRGWRRSGKYVYKPVMNQTCCPQYTIRCRPLQFQPS | |
RUO | |
ATE1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
11101 | |
ATE1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC26724 | |
ATE1 | |
Recombinant | |
wheat germ expression system |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction