Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ATP11B Partial ORF (NP_055431, 1087 a.a. - 1177 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00023200Q01S

 View more versions of this product

Catalog No. 89-935-985


Add to cart

Description

Description

P-type ATPases, such as ATP11B, are phosphorylated in their intermediate state and drive uphill transport of ions across membranes. Several subfamilies of P-type ATPases have been identified. One subfamily transports heavy metal ions, such as Cu(2+) or Cd(2+). Another subfamily transports non-heavy metal ions, such as H(+), Na(+), K(+), or Ca(+). A third subfamily transports amphipaths, such as phosphatidylserine.[supplied by OMIM]

Sequence: DIIKKVFDRHLHPTSTEKAQLTETNAGIKCLDSMCCFPEGEAACASVGRMLERVIGRCSPTHISRSWSASDPFYTNDRSILTLSTMDSSTC
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.75kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DIIKKVFDRHLHPTSTEKAQLTETNAGIKCLDSMCCFPEGEAACASVGRMLERVIGRCSPTHISRSWSASDPFYTNDRSILTLSTMDSSTC
RUO
ATP11B
Wheat Germ (in vitro)
GST
NP_055431
Liquid
23200
ATP11B (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ATPIF/ATPIR/DKFZp434J238/DKFZp434N1615/KIAA0956/MGC46576
ATP11B
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit