Learn More
Abnova™ Human ATP11B Partial ORF (NP_055431, 1087 a.a. - 1177 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_055431 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 23200 |
Molecular Weight (g/mol) | 35.75kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-935-986
|
Abnova™
H00023200Q01L |
25 ug |
Each for $769.00
|
|
|||||
89-935-985
|
Abnova™
H00023200Q01S |
10 ug |
Each for $499.00
|
|
|||||
Description
P-type ATPases, such as ATP11B, are phosphorylated in their intermediate state and drive uphill transport of ions across membranes. Several subfamilies of P-type ATPases have been identified. One subfamily transports heavy metal ions, such as Cu(2+) or Cd(2+). Another subfamily transports non-heavy metal ions, such as H(+), Na(+), K(+), or Ca(+). A third subfamily transports amphipaths, such as phosphatidylserine.[supplied by OMIM]
Sequence: DIIKKVFDRHLHPTSTEKAQLTETNAGIKCLDSMCCFPEGEAACASVGRMLERVIGRCSPTHISRSWSASDPFYTNDRSILTLSTMDSSTCSpecifications
NP_055431 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.75kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATPIF/ATPIR/DKFZp434J238/DKFZp434N1615/KIAA0956/MGC46576 | |
ATP11B | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
23200 | |
ATP11B (Human) Recombinant Protein (Q01) | |
DIIKKVFDRHLHPTSTEKAQLTETNAGIKCLDSMCCFPEGEAACASVGRMLERVIGRCSPTHISRSWSASDPFYTNDRSILTLSTMDSSTC | |
RUO | |
ATP11B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.